missing translation for 'onlineSavingsMsg'
Learn More

Meditrade™ Gentle Skin™ Grip Latex Examination Gloves, PPE Cat. III

Product Code. 17103911
Change view
Click to view available options
Size:
Medium
X-Large
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17103911 X-Large Pack of 100
17183901 Medium Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17103911 Supplier Meditrade™ Supplier No. 1221GRIPXL

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Meditrade™ disposable, non-sterile latex examination gloves are textured to provide an excellent grip. The ambidextrous gloves are natural white color with a beaded cuff.

  • AQL 1.5
  • Class I Medical
  • PPE Category III
  • EN ISO 13485:2016
  • Food compliant
  • Gentle Skin™ grip
  • Latex with polymer coated surface
  • Natural white
  • Powder-free
  • Non-sterile
  • Textured
  • Beaded cuff
  • Ambidextrous
  • Disposable
  • Five year shelf life

Specifications

Certifications/Compliance AQL 1.5
Certifications/Compliance (Europe) EN 455-1/2/3/4, EN 374-1/2/4/5, EN 16523-1, EN 420, EN ISO 13485:2016, Food Compliant
Color Natural White
Disposable Single-use
Laboratory/Cleanroom Use Laboratory
Latex Free Yes
Material Latex
Size X-Large
Thickness (Metric) 0.080 mm (Cuff)
Series 1221 Grip
Length (Metric) 240 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.