missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Geranic acid, 98% (sum of isomers)
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.955 |
| Boiling Point | 249°C to 251°C |
| Flash Point | 133°C (271°F) |
| Linear Formula | (CH3)2C=CHCH2CH2C(CH3)=CHCO2H |
| Refractive Index | 1.484 |
| Quantity | 100 g |
| Beilstein | 1763804 |
| Formula Weight | 168.24 |
| Percent Purity | ≥90% |
| Chemical Name or Material | Geranic acid, sum of isomers |
Safety and Handling
Product Identifier
- Geranic acid
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Geranic acid
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction