missing translation for 'onlineSavingsMsg'
Learn More

Geraniol, 97%

Product Code. 11454764
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11454764

Brand: Thermo Scientific Alfa Aesar A13736.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 106-24-1 | C10H18O | 154.253 g/mol

Geraniol is used in the synthesis of insect repellant. It is also used in the synthesis of Angelicoin A and Herecinone J, which inhibit collagen-induced platelet aggregation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Geraniol is used in the synthesis of insect repellant. It is also used in the synthesis of Angelicoin A and Herecinone J, which inhibit collagen-induced platelet aggregation.

Solubility
Not miscible in water.

Notes
Incompatible with Strong oxidizing agents, acid chlorides, acid anhydrides. Keep away from heat. Air sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-24-1
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00002917
InChI Key GLZPCOQZEFWAFX-JXMROGBWSA-N
Synonym geraniol, lemonol, geranyl alcohol, trans-geraniol, e-geraniol, e-nerol, trans-3,7-dimethyl-2,6-octadien-1-ol, e-3,7-dimethylocta-2,6-dien-1-ol, 2e-3,7-dimethylocta-2,6-dien-1-ol, geraniol extra
PubChem CID 637566
ChEBI CHEBI:17447
IUPAC Name (2E)-3,7-dimethylocta-2,6-dien-1-ol
SMILES CC(=CCCC(=CCO)C)C

Specifications

Melting Point -15°C
Density 0.884
Boiling Point 114°C to 115°C (12 mmHg)
Flash Point 101°C (213°F)
Odor Floral
Refractive Index 1.476
Quantity 250 g
Beilstein 1722456
Merck Index 14,4403
Solubility Information Not miscible in water.
Formula Weight 154.25
Percent Purity 97%
Chemical Name or Material Geraniol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Geraniol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Geraniol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.