missing translation for 'onlineSavingsMsg'
Learn More

Geraniol, 99%

Product Code. 10368813 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Drum
Unit Size:
100g
2.5kg
25g
500g
5g
Product Code. Quantity Packaging unitSize
10368813 500 g Glass bottle 500g
10314702 5 g Glass bottle 5g
10229840 25 g Glass bottle 25g
10509092 100 g Glass bottle 100g
15189283 2.5 kg Plastic Drum 2.5kg
5 options
This item is not returnable. View return policy

Product Code. 10368813

Brand: Thermo Scientific Acros 410905000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-24-1 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-24-1
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00002917
InChI Key GLZPCOQZEFWAFX-JXMROGBWSA-N
Synonym geraniol, lemonol, geranyl alcohol, trans-geraniol, e-geraniol, e-nerol, trans-3,7-dimethyl-2,6-octadien-1-ol, e-3,7-dimethylocta-2,6-dien-1-ol, 2e-3,7-dimethylocta-2,6-dien-1-ol, geraniol extra
PubChem CID 637566
ChEBI CHEBI:17447
IUPAC Name (2E)-3,7-dimethylocta-2,6-dien-1-ol
SMILES CC(=CCCC(=CCO)C)C

Specifications

Melting Point -15.0°C
Color Colorless to Yellow
Density 0.8780g/mL
Boiling Point 229.0°C to 230.0°C (757.0 mmHg)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C=CHCH2CH2C(CH3)=CHCH2OH
Refractive Index 1.4750 to 1.4790
Quantity 500 g
Beilstein 01,457
Merck Index 15,4437
Specific Gravity 0.878
Solubility Information Solubility in water: practically insoluble. Other solubilities: miscible with alcohol and ether, ethanol 50 vol. % (20°C):1:10 v:v
Formula Weight 154.25
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Geraniol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.