missing translation for 'onlineSavingsMsg'
Learn More

Geraniol, TRC

Product Code. 30137083
Change view
Click to view available options
Quantity:
5 g
10 g
250 g
Unit Size:
10g
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137083 250 g 250g
30136276 10 g 10g
17914445 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137083 Supplier Toronto Research Chemicals Supplier No. TRCG367000250G
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-24-1
Molecular Formula C10 H18 O
Molecular Weight (g/mol) 154.25
Synonym 2,6-Octadien-1-ol, 3,7-dimethyl-, (2E)-, 2,6-Octadien-1-ol, 3,7-dimethyl-, (E)- (8CI), (2E)-3,7-Dimethyl-2,6-octadien-1-ol, (E)-3,7-Dimethyl-2,6-octadien-1-ol, (E)-3,7-Dimethyl-2,6-octadienol, (E)-Geraniol, (E)-Nerol, 3,7-Dimethyl-trans-2,6-octadien-1-ol, Geraniol, Geraniol 60, Geraniol 980, Geraniol BJ, Geranyl alcohol, Lemonol, MosquitoSafe, NSC 9279, TT 4228, TT 4302, trans-1-Hydroxy-3,7-dimethyl-2,6-octadiene, trans-3,7-Dimethyl-2,6-octadien-1-ol, trans-Geraniol, β-Geraniol
IUPAC Name (2E)-3,7-dimethylocta-2,6-dien-1-ol
SMILES CC(=CCC\C(=C\CO)\C)C

Specifications

Quantity 250 g
InChI Formula InChI=1S/C10H18O/c1-9(2)5-4-6-10(3)7-8-11/h5,7,11H,4,6,8H2,1-3H3/b10-7+
Formula Weight 154.1358
Chemical Name or Material Geraniol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.