missing translation for 'onlineSavingsMsg'
Learn More

Geranyl bromide, 96%

Product Code. 11478036
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478036 5 g 5g
11488036 25 g 25g
11498036 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478036 Supplier Thermo Scientific Alfa Aesar Supplier No. A14093.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6138-90-5 | C10H17Br | 217.15 g/mol

Geranyl bromide is used for the preparation of 3,7-dihydroxyflavone derivatives and baicalein. It is also involved in the synthesis of potential flavonoidic modulators of P-glycoprotein activity. Further, it reacts with benzenesulfinic acid to prepare geranyl phenyl sulfone. In addition to this, it takes part in the palladium catalyzed cross coupling with aryl and alkenylgold(I) phosphanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Geranyl bromide is used for the preparation of 3,7-dihydroxyflavone derivatives and baicalein. It is also involved in the synthesis of potential flavonoidic modulators of P-glycoprotein activity. Further, it reacts with benzenesulfinic acid to prepare geranyl phenyl sulfone. In addition to this, it takes part in the palladium catalyzed cross coupling with aryl and alkenylgold(I) phosphanes.

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6138-90-5
Molecular Formula C10H17Br
Molecular Weight (g/mol) 217.15
MDL Number MFCD00000243
InChI Key JSCUZAYKVZXKQE-JXMROGBWSA-N
Synonym geranyl bromide, e-1-bromo-3,7-dimethylocta-2,6-diene, 2,6-octadiene, 1-bromo-3,7-dimethyl-, e, 2,6-octadiene, 1-bromo-3,7-dimethyl-, 2e, geranylbromide, 2e-1-bromo-3,7-dimethyl-2,6-octadiene, 6e-8-bromo-2,6-dimethylocta-2,6-diene, 1-bromo-3,7-dimethyl-2,6-octadiene, trans-geranyl bromide, e-geranyl bromide
PubChem CID 5365867
IUPAC Name (2E)-1-bromo-3,7-dimethylocta-2,6-diene
SMILES CC(=CCCC(=CCBr)C)C

Specifications

Density 1.094
Boiling Point 99°C to 101°C (10 mmHg)
Flash Point 95°C (203°F)
Refractive Index 1.504
Quantity 5 g
Beilstein 1422412
Sensitivity Light sensitive
Solubility Information Immiscible with water.
Formula Weight 217.16
Percent Purity 96%
Chemical Name or Material Geranyl bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Geranyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1-bromo-3,7-dimethyl-2,6-Octadiene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.