missing translation for 'onlineSavingsMsg'
Learn More

Geranyl linalool, ca. 95%, Tech., mixture of isomers

Product Code. 10500214 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
25 g
Unit Size:
2.5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10500214 2.5 g 2.5g
10307801 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10500214 Supplier Thermo Scientific Acros Supplier No. 278360025
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1113-21-9 | C20H34O | 290.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1113-21-9
Molecular Formula C20H34O
Molecular Weight (g/mol) 290.49
MDL Number MFCD00059363
InChI Key IQDXAJNQKSIPGB-HQSZAHFGNA-N
Synonym geranyl linalool, geranyllinalool, 3,7,11,15-tetramethyl-1,6,10,14-hexadecatetraen-3-ol, e,e-geranyllinalool, 6e,10e-geranyllinalool, 3,7,11,15-tetramethylhexadeca-1,6,10,14-tetraen-3-ol, 6e,10e-3,7,11,15-tetramethylhexadeca-1,6,10,14-tetraen-3-ol, e,e-3,7,11,15-tetramethylhexadeca-1,6,10,14-tetraen-3-ol, 1,6,10,14-hexadecatetraen-3-ol, 3,7,11,15-tetramethyl-, 6e,10e, 1,6,10,14-hexadecatetraen-3-ol, 3,7,11,15-tetramethyl
PubChem CID 5365872
ChEBI CHEBI:74299
IUPAC Name (6E,10E)-3,7,11,15-tetramethylhexadeca-1,6,10,14-tetraen-3-ol
SMILES CC(C)=CCC\C(C)=C\CC\C(C)=C\CCC(C)(O)C=C

Specifications

Color Colorless to Yellow
Density 0.8800g/mL
Boiling Point 250.0°C
Flash Point 160°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4860 to 1.4920
Quantity 2.5 g
Specific Gravity 0.88
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents, solubel in 70% ethanol
Formula Weight 290.48
Percent Purity ca. 95%
Grade Technical
Physical Form Liquid
Chemical Name or Material Geranyl linalool, tech., ca. 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.