missing translation for 'onlineSavingsMsg'
Learn More

Geranyl Linalool, TRC

Product Code. 30240654
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240654 1 g 1g
30240497 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30240654 Supplier Toronto Research Chemicals Supplier No. TRCG3677501G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1113-21-9
Molecular Formula C20H34O
Molecular Weight (g/mol) 290.48
Synonym 3,7,11,15-Tetramethyl-1,6,10,14-hexadecatetraen-3-ol, 3,7,11,15-Tetramethyl-1,6,10,14-cetene-3-ol, 3,7,11,15-Tetramethyl-3-hydroxy-1,6,10,14-hexadecatetraene;
IUPAC Name (6E,10E)-3,7,11,15-tetramethylhexadeca-1,6,10,14-tetraen-3-ol
SMILES CC(=CCC\C(=C\CC\C(=C\CCC(C)(O)C=C)\C)\C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H34O/c1-7-20(6,21)16-10-15-19(5)14-9-13-18(4)12-8-11-17(2)3/h7,11,13,15,21H,1,8-10,12,14,16H2,2-6H3/b18-13+,19-15+
Formula Weight 290.261
Chemical Name or Material Geranyl Linalool
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.