missing translation for 'onlineSavingsMsg'
Learn More

Germanium(IV) methoxide, 98%

Product Code. 11498357
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498357 25 g 25g
11488357 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11498357 Supplier Thermo Scientific Alfa Aesar Supplier No. A18437.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 992-91-6 | C4H12GeO4

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 992-91-6
Molecular Formula C4H12GeO4
MDL Number MFCD00014881
Synonym tetramethoxygermanium, tetramethoxygermanium iv, methyl germanate iv

Specifikationer

Boiling Point 145°C
Melting Point -18°C
Quantity 25 g
Flash Point 40°C (105°F)
Odor Like Alcohol
Refractive Index 1.4015
Sensitivity Moisture sensitive
Density 1.3254^2^0
Linear Formula (CH3O)4Ge
UN Number UN2929
Formula Weight 196.73
Percent Purity 98%
Chemical Name or Material Germanium(IV) methoxide
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.