missing translation for 'onlineSavingsMsg'
Learn More

Germanium(IV) oxide, 99.99% (metals basis)

Product Code. 11387828
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11387828

Brand: Thermo Scientific Alfa Aesar 036382.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in Phosphors, Transistors and Diodes, and Infrared Transmitting Glass | CAS: 1310-53-8 | GeO2 | 104.628 g/mol

Used in phosphors, transistors and diodes, and infrared transmitting glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-53-8
Molecular Formula GeO2
Molecular Weight (g/mol) 104.628
MDL Number MFCD00011030
InChI Key YBMRDBCBODYGJE-UHFFFAOYSA-N
Synonym germanium dioxide, germania, germanic acid, germanium oxide, germanium iv oxide, germanic oxide crystalline, dioxogermanium, germanic oxide, wln: ge o2
PubChem CID 14796
IUPAC Name dioxogermane
SMILES O=[Ge]=O

Specifications

Melting Point 1086°C
Color White
Physical Form Sintered Lump
Quantity 10 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information soluble in water; insoluble in HF and HCl.
Formula Weight 104.59
Odor Odorless
Density 6.239
Chemical Name or Material Germanium(IV) oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.