missing translation for 'onlineSavingsMsg'
Learn More

Germanium pieces, 2cm (0.8in) & down, 99.9999% (metals basis)

Product Code. 11340669
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11340669 50 g 50g
11350669 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11340669 Supplier Thermo Scientific Alfa Aesar Supplier No. 000123.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7440-56-4 | Ge | 72.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-56-4
Molecular Formula Ge
Molecular Weight (g/mol) 72.63
MDL Number MFCD00085310
InChI Key GNPVGFCGXDBREM-UHFFFAOYSA-N
Synonym element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder
PubChem CID 6326954
ChEBI CHEBI:30441
IUPAC Name germanium
SMILES [Ge]

Specifications

Quantity 50 g
Odor Odorless
Particle Size 2cm (0.8 in.) & down
Form Pieces
Percent Purity 99.9999%
Assay (metals basis)
Chemical Name or Material Germanium pieces, Electrical Resistivity: 40 ohm-cm

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.