missing translation for 'onlineSavingsMsg'
Learn More

Vilber Lourmat™ Germicidal UV Lamp

Product Code. 11880370
Change view
Click to view available options
Length (Metric):
500 mm
505 mm
980 mm
1290 mm
Wattage:
15 W
30 W
45 W
60 W
80 W
120 W
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Wattage Unit Size
11880370 500 mm 30 W Each
10262400 505 mm 15 W Each
11746768 980 mm 30 W Each
11706778 980 mm 60 W Each
11766728 500 mm 45 W Each
11716678 1290 mm 80 W Each
11766778 1290 mm 120 W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 11880370 Supplier Vilber Lourmat™ Supplier No. 312121511
  • ONLINE EXCLUSIVE PRICE
    1199.74 €
    498.86 € / Pack of 1
    Save 700.88 €
    58% Off
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Voltage 230 V
Wattage 30 W
Wavelength 254 nm
Depth (Metric) 140 mm
Electrical Requirements 30 W, 230 V, 50 Hz
Height (Metric) 110 mm
Length (Metric) 500 mm
Lighting Intensity 76 mW/cm2
Packaging Quantity 1 Ea.
Product Type Germicidal UV Lamp
Frequency 50 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.