missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ GF 109203X

Product Code. 30002170
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30002170 10 mg 10mg
18025692 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30002170 Supplier R&D Systems™ Supplier No. 0741/10
  • ONLINE EXCLUSIVE PRICE
    298.75 €
    269.85 € / 10mg
    Save 28.90 €
    9% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Protein kinase C inhibitor

Very potent and selective inhibitor of protein kinase C, selective for the α and β1 isoforms (IC50 values are 0.0084, 0.0180, 0.210, 0.132, and 5.8 μM for α, β1, δ, ε and ζ isoforms respectively). Selective over MLCK, PKG and PKA (IC50 values are 0.6, 4.6, and 33 μM respectively). Potent antagonist at the 5-HT3 receptor (Ki = 29.5 nM). Anti-inflammatory in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133052-90-1
Molecular Formula C25H24N4O2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 412.49
Purity >98%
Molecular Weight (g/mol) 412.493
Chemical Name or Material GF 109203X
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.