Learn More
Aalborg™ TIO-GFM Mounting Kit
Product Code. 13594500
Change view
Click to view available options
Type:
GFM Mass Flow Meter
Unit Size:
Each
Product Code. Type Unit Size
13594500 GFM Mass Flow Meter Each
1 options
Product Code. 13594500 Supplier Aalborg™ Supplier No. KITTMDD
  • ONLINE EXCLUSIVE PRICE
    81.90 €
    49.98 € / Pack of 1
    Save 31.92 €
    38% Off
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed especially for the laboratory these kits offer a diverse economical way of acquiring flow measurement capabilities. Aalborg™ TIO-GFM Mounting Kit allows for easy changing of flow tubes in the field.
  • Kits are shipped in convenient molded plastic carrying cases.
  • Flow tubes and floats are interchangeable in frames supplied facilitating overlapping flow ranges.
  • For non-corrosive fluids use the Aluminum Kit.
  • For corrosives consider the Stainless Steel Kit.
  • For corrosive applications at lower pressure or for high purity fluid service specify the PTFE Kit.
  • Supplied with glass floats installed.
Specifications
Type GFM Mass Flow Meter
For Use With (Application) Measures flow rate