missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GFP-EIF2A

Product Code. 10003153
Change view
Click to view available options
Quantity:
20 nmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10003153 20 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10003153 Supplier Invitrogen™ Supplier No. PV4809
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

GFP-EIF2A

EIF2A has been expressed as a fusion with GFP (Green Fluorescent Protein) to act as a physiological substrate for the EIF2AK family of kinases including PERK (EIF2AK3) and PKR (EIF2AK2).

For use in combination with the LanthaScreen™ Tb-anti-eIF2alpha [pSer52] Antibody (PV4810) to evaluate inhibitors in medium to high throughput screening applications.

TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Biochemical terbium phospho-specific activity
Content And Storage GFP-EIF2alpha protein substrate in 9 mM HEPES pH 7.5, 123 mM NaCl, 2.43 mM KCl, 10% Glycerol, 2mM DTT . Store at -80 °C.
Detection Method Fluorescence
Form Liquid
Quantity 20 nmol
Substrate EIF2A
Substrate Properties Protein-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Serine/Threonine Kinases
Gene ID (Entrez) 9833
Label or Dye GFP (EmGFP)
Molecular Weight (g/mol) 69100 Da
Shipping Condition Dry Ice
Target Entry EIF2AK1, EIF2AK2, EIF2AK3
Validated Kinases EIF2AK2, EIF2AK3
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.