Learn More
IBA Lifesciences GFP-Strep-tag control protein
GFP-Strep-tag control protein for Strep-Tactin™ and Strep-Tactin™XT applications
Brand:  IBA Lifesciences 2-1007-005
Additional Details : Weight : 0.00050kg
 View more versions of this product
Product Code. 15503077
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • GFP control protein consisting of the green fluorescent protein (GFP) C-terminally tagged with the Strep-tag™II or Twin-Strep-tag™.
  • It is suitable for test purifications, detections, and immobilizations with Strep-Tactin™ or Strep-Tactin™XT.  
Specifications
Specifications
0.5 mL
500 μg of a 30 kDa protein
Twin-Strep-tag™
Product Suggestions
Product Suggestions
Videos
SDS
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote
For Research Use Only.