Learn More
IBA Lifesciences GFP-Strep-tag control protein
GFP-Strep-tag control protein for Strep-Tactin™ and Strep-Tactin™XT applications
104.00 € - 120.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15593067
|
IBA Lifesciences
2-1006-005 |
5 mL |
120.00 €
500µg |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
15503077
|
IBA Lifesciences
2-1007-005 |
0.5 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- GFP control protein consisting of the green fluorescent protein (GFP) C-terminally tagged with the Strep-tag™II or Twin-Strep-tag™.
- It is suitable for test purifications, detections, and immobilizations with Strep-Tactin™ or Strep-Tactin™XT.
Specifications
500 μg of a 30 kDa protein |
For Research Use Only.