Learn More
GFR alpha-1/GDNF R alpha-1 Goat anti-Rat, Polyclonal, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 21 publication
Brand:  R&D Systems AF560-SP
Encompass
Product Code. 15748732
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
GFR alpha-1/GDNF R alpha-1 Polyclonal specifically detects GFR alpha-1/GDNF R alpha-1 in Rat samples. It is validated for Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction.