Learn More
Alfa Aesar™ GGTI-2147
Brand: Alfa Aesar™ J65022.|D
Code : NEW
Additional Details : CAS Number : 191102-87-1 Weight : 0.00001kg
Specifications
0.5 mg | |
-30°C to -10°C | |
191102-87-1 | |
470.56 | |
Light Sensitive and Hygroscopic | |
GGTI-2147 |
Off-white | |
A cell-permeable non-thiol peptidomimetic that acts as a potent and selective inhibitor of geranylgeranyltransferase I | |
C28H30N4O3 | |
(S)-Methyl 2-(4-(((1H-imidazol-4-yl)methyl)amino)-2-(naphthalen-1-yl)benzamido)-4-methylpentanoate; | |
Soluble in DMSO | |
Solid |