missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Giant Stir Bar

Product Code. 10379640 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
57 mm
80 mm
108 mm
110 mm
136 mm
159 mm
Diameter (Metric):
14 mm
24 mm
27 mm
36 mm
Shape:
Cylinder
Cylinder, Pivot ring
Triangle
Unit Size:
Each
Pack of 5
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Shape unitSize
10379640 108 mm 24 mm Cylinder, Pivot ring Each
10074400 159 mm 24 mm Cylinder, Pivot ring Each
10410211 57 mm 24 mm Cylinder, Pivot ring Each
10166330 108 mm 27 mm Cylinder Each
10369640 159 mm 27 mm Cylinder Each
10470011 57 mm 27 mm Cylinder Each
11868932 80 mm 14 mm Triangle Pack of 5
10480971 80 mm 14 mm Triangle Each
10688071 110 mm 36 mm Triangle Each
12706108 136 mm 36 mm Triangle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 10379640
Spotlight Offers
Use promo code 28036
View all promotions
In Stock
Add to basket
Edge
Add to basket

Designed for use in large containers and vials. These giant PTFE-coated magnetic stir bares are autoclavable.

  • Stable, general-purpose magnetic stir bar
  • Designed for use in large containers and vials
  • Autoclavable

Specifications

Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core; Cylindrical with Pivot Ring
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Diameter (Metric) 24 mm
Length (Metric) 108 mm
Quantity 1/Ea.
Shape Cylinder, Pivot ring
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.