Learn More
Gibberellic acid, 90+%
Description
It is a plant growth regulating hormone that promotes plant cell growth and elongation. They promote rapid stem and root growth, induce mitotic division and initiate (break dormancy) and increase seed germination rates. They are also involved in processes such as gravitropism, tensioning and floral display. Gibberellin A3 (G1025, G7645) and A4 (G7276) may be used as supplements to plant growth media such as Murashige and Skoog media.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is a plant growth regulating hormone that promotes plant cell growth and elongation. They promote rapid stem and root growth, induce mitotic division and initiate (break dormancy) and increase seed germination rates. They are also involved in processes such as gravitropism, tensioning and floral display. Gibberellin A3 (G1025, G7645) and A4 (G7276) may be used as supplements to plant growth media such as Murashige and Skoog media.
Solubility
Soluble in water 20°C 5g/L. Soluble in Ethanol: 200 mg/4ml.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 222°C (decomposition) |
| Quantity | 5 g |
| Beilstein | 54346 |
| Merck Index | 14,4419 |
| Solubility Information | Soluble in water 20°C 5g/L. Soluble in Ethanol: 200mg/4ml. |
| Optical Rotation | +78° (c=2 in Methanol) |
| Formula Weight | 346.37 |
| Percent Purity | ≥90% |
| Chemical Name or Material | Gibberellic acid |
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.