missing translation for 'onlineSavingsMsg'
Learn More

Gibberellic Acid, 90%, TRC

Product Code. 30133210
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133210 500 mg 500mg
30135889 1 g 1g
17952005 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133210 Supplier Toronto Research Chemicals Supplier No. TRCG377450500MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-06-5
Molecular Formula C19 H22 O6
Molecular Weight (g/mol) 346.37
Synonym Gibb-3-ene-1,10-dicarboxylic acid, 2,4a,7-trihydroxy-1-methyl-8-methylene-, 1,4a-lactone, (1α,2β,4aα,4bβ,10β)-, Gibberellic acid (7CI,8CI), Gibberellin A3 (6CI), (+)-Gibberellic acid, (+)-Gibberellin A3, Activol, Arbostim, Berelex, Berelex L, Clemencuaje, GA, GA3, GibGro, Gibberellic acid GA3, Gibberellin GA3, Giberllon, Gibrescol, Gibreskol, Grocel GA3, Maxon, Maxon (plant growth regulator), NSC 14190, NSC 19450, Pro-Gibb, Pro-Gibb Plus, ProGib, Ralex, Release, Release LC, RyzUp, RyzUp SmartGrass, TU 64-3-103-75
IUPAC Name (3S,3aS,4S,4aS,6S,8aR,8bR,11S)-6,11-Dihydroxy-3-methyl-12-methylene-2-oxo-4a,6-ethano-3,8b-prop-1-enoperhydroindeno[1,2-b]furan-4-carboxylic acid
SMILES C[C@]12[C@@H](O)C=C[C@@]3(OC1=O)[C@@H]4CC[C@]5(O)C[C@]4(CC5=C)[C@H]([C@H]23)C(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C19H22O6/c1-9-7-17-8-18(9,24)5-3-10(17)19-6-4-11(20)16(2,15(23)25-19)13(19)12(17)14(21)22/h4,6,10-13,20,24H,1,3,5,7-8H2,2H3,(H,21,22)/t10-,11+,12-,13-,16-,17+,18+,19-/m1/s1
Formula Weight 346.1416
Percent Purity >95%
Chemical Name or Material Gibberellic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.