missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AmnioMAX™ C-100 Supplement

Product Code. 10772077 Shop All Gibco Products
Change view
Click to view available options
Quantity:
15 mL
75 mL
Unit Size:
75mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10772077 15 mL Each
10507485 75 mL 75mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10772077 Supplier Gibco™ Supplier No. 12556015
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A part of kit used to prepare fully supplemented Gibco AmnioMAX C-100 Complete Medium developed for short term culture of human amniotic fluid cells for cytogenetic studies and in vitro diagnostic procedures

Gibco™ AmnioMAX™ C-100 Supplement is part of a kit used to prepare a fully-supplemented Gibco™ AmnioMAX™ C-100 Complete Medium developed for the short term culture of human amniotic fluid cells for cytogenetic studies and in vitro diagnostic procedures.

Gibco™ AmnioMAX™ C-100 Complete Medium features:

  • Easy-to-use format
  • Quality and performance testing
  • Unique, optimized formulation

Easy-to-use format

Gibco™ AmnioMAX™ C-100 Complete Medium is a two-part kit for preparation of a complete medium that requires no additional supplementation. The complete medium can be stored at 2–8°C for up to ten days.

Quality and performance testing

Every lot of Gibco™ AmnioMAX™ C-100 Complete Medium is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance. Pooled primary human amniotic fluid samples are cultured for six days in Gibco™ AmnioMAX™ C-100 Complete Medium before measuring the total number of colonies and total number of mitotic colonies. In addition, each lot is tested for pH and osmolality and must pass a sterility test prior to lot release.

Unique, optimized formulation

Gibco™ AmnioMAX™ C-100 Complete Medium contains Fetal Bovine Serum (FBS), gentamicin, and L-glutamine to maximize cell attachment and growth. This optimized medium also has an enhanced buffering system that provides greater pH stability during culture manipulations.

Product Use

For in vitro diagnostic use.

cGMP Manufacturing and Quality System

Gibco™ AmnioMAX™ C-100 Supplement is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Order Info

Shipping Condition: Dry Ice

Compliance

Gibco AmnioMAX C-100 Supplement is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: -5 to -20°C
Shipping conditions: Frozen
Shelf life: 16 months from date of manufacture. Use complete medium within 7–10 days once opened.
Form Frozen
Product Type C-100 Supplement
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Glutamine
Product Line AmnioMAX
Quantity 15 mL
Shipping Condition Dry Ice

In vitro diagnostic

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.