missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Breast Cancer Cell Culture Starter Kit, Europe

Product Code. 16669522
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16669522 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16669522 Supplier Gibco™ Supplier No. A3518401

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provides researchers with a convenient package of reagents for the culture and transfection of cancer cell types frequently used in cancer research.

  • Gibco Cancer Cell Culture Starter kits provide researchers with a convenient package of reagents for the culture and transfection of cancer cell types frequently used in cancer research. Cancer Cell Culture Starter kits are currently available for breast, lung, prostate, and liver cancer cells.
  • Each starter kit includes the following products for cancer cell culture and transfection: Basal media, Fetal bovine serum, Penicillin-streptomycin, TrypLE Express Enzyme, Lipofectamine 3000 Transfection Reagent, Opti-MEM I
  • The Cancer Cell Culture Starter kits contain the most commonly used reagents to help you start your cancer cell culture quickly and achieve successful transfections, based on a review of hundreds of cancer research publications in which the following cell lines were used: Prostate cancer: DU145, PC-3, LNCaP, Lung cancer: A549, NCI-H975, PC-14, NCI-H1299, Breast cancer: MCF-7, MDA-MB-231, T-47D, BT-474, Liver cancer: Hep G2, Hep 3B
  • The Cancer Culture Starter kits also include a suggested protocol for the culture, dissociation (cell passage), and transfection of the most-published cancer cell lines. Achieve peace of mind and quickly initiate your experiments with these starter kits, the components of which may also be purchased separately.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Cancer Cells
Product Type Culture Starter Kit
Sterility Sterile-filtered
For Use With (Application) Mammalian Cell Culture
Quantity 1 Kit
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Format Kit
Content And Storage • DMEM, high glucose, with GlutaMAX Supplement (store at 2–8°C)
• Fetal bovine serum, qualified, One Shot™ format, EU Approved South America origin (store at -5 to -30°C)
• Penicillin-streptomycin (10,000 U/mL) (store at -5 to -30°C)
• TrypLE Express Enzyme (1X), no phenol red (store at room temperature)
• Lipofectamine 3000 Transfection Reagent (store at 2–8°C)
• Opti-MEM I Reduced Serum Medium (store at 2–8°C)

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.