Learn More
Description
Contains (1X) containing calf serum as a trypsin inhibitor.
Sustainable packaging
Cell Culture, Corneal Epithelial Cell Culture, Fibroblast Culture, Keratinocyte Culture, Large Vessel Endothelial Cell Culture, Mammalian Cell Culture, Mammary Epithelial Cell Culture, Melanocyte Culture, Microvascular Endothelial Cell Culture, Primary Cell Culture, Smooth Muscle Cell Culture
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | minus 20 degrees C |
| Cell Type | Mammalian Cells |
| Concentration | 1 X |
| Product Type | Trypsin inhibitor |
| Sterility | Sterile-filtered |
| pH | 7.4 |
| Product Line | Cascade Biologics |
| Quantity | 100 mL |
| Shipping Condition | Room Temperature |
Frequently Asked Questions (FAQs)
Please use this selection chart that compares our cell dissociation reagents (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/reagents/trypsin.html).
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
