missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Collagenase, Type I, powder

Product Code. 10114532
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10114532 1 g 1g
11500536 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10114532 Supplier Gibco™ Supplier No. 17100017

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cleaves bond between neutral amino acid (X) and glycine in sequence Pro-X-Gly-Pro, which is found with high frequency in collagen

  • Collagenase is a protease that cleaves the bond between a neutral amino acid (X) and glycine in the sequence Pro-X-Gly-Pro, which is found with high frequency in collagen
  • It is unique among proteases in its ability to degrade the triple-helical native collagen fibrils commonly found in connective tissues such as skin, tendon, blood vessels, and bone
  • Collagenase disaggregation is suitable for the culture of human tumors, mouse kidney, human adult and fetal brain, and many other tissues including epithelium
  • Collagenase is relatively gentle, dissociates well at physiological temperature and pH, and requires no mechanical agitation or special equipment
  • Gibco™ Collagenase Type I is isolated from Clostridium histolyticum and packaged as a lyophilized, non-sterile powder for research use in cell or tissue dissociation, and organ perfusions
  • Compared to other collagenase preparations, Gibco™ Collagenase Type I has average levels of collagenase, caseinase, clostripain, and tryptic activities, and is well-suited for the digestion of fat, adrenal, and liver cells or tissues
  • Gibco™ Collagenase Type I activity is guaranteed to be greater than 125 units/mg
  • Store at 2°C to 8°C, protected from light

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2°C to 8°C. Protect from light.
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture
Form Lyophilized
Product Type Collagenase
Sterility Non-sterile
Quantity 1 g
Shipping Condition Room Temperature
Can collagenase be used to dissociate feeder free hESC when cultured on CELLstart medium?

Yes, collagenase can be used. It is recommended to use a 10 mg/ml collagenase IV solution (0.5 ml per 6 cm dish). After 1 minute, remove collagenase and rinse cells. Scrape cells into wash buffer (see product manual for more information), pellet, and re-plate as desired.

How do I make the 1000X stock (100 U/µL) solution from Collagenase powder?

1. Add 1 mL Hanks' Balanced Salt Solution (HBSS) with calcium and magnesium directly to 1 g vial of Collagenase. Vortex gently to ensure complete dissolution. Transfer to a clean tube.
2. Determine volume of HBSS (with calcium and magnesium) required to bring collagenase solution to 100 U/µL (1000X stock solution). The activity is lot- specific. Rinse vial with this volume of HBSS (with calcium and magnesium), and combine. Filter sterilize 1000X stock solution with a low protein binding filtration unit.
Example: Assuming the lot you have purchased has an activity of 265 U/mg, this lot will have 265000 Units per mL when you reconstitute collagenase into HBSS (with calcium and magnesium) at 1 g/mL. In order to dilute 265000 U/L to 100000 U/mL (= 100 U/µL), you need to dilute the 1 g/mL enzyme solution 2.65 fold.

What reagents do you offer for cell dissociation, and what are the main differences between them?

Please use this selection chart that compares our cell dissociation reagents (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/reagents/trypsin.html).

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.