missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Dynabeads™ Human T-Activator CD3/CD28/CD137

Product Code. 10573269
Change view
Click to view available options
Volume (Metric):
0.4 mL
2 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10573269 2 mL Each
10142878 0.4 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10573269 Supplier Gibco™ Supplier No. 11163D

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Includes

0.4mL DynabeadsHuman T-Activator CD3/CD28/CD137 is supplied as a suspension containing 1 x 105 Dynabeads/mL in phosphate buffered saline (PBS), pH 7.4, w/ 0.1% human serum albumin (HSA)

Dynabeads Human T-Activator CD3/CD28/CD137 beads are 4.5-μm superparamagnetic beads covalently coupled with an optimized combination of anti-human CD3, CD28, and CD137 antibodies and intended for ex vivo expansion/proliferation of human antigen (Ag)-specific T cells.

Dynabeads Human T-Activator CD3/CD28/CD137 beads are 4.5-μm superparamagnetic beads covalently coupled with an optimized combination of anti-human CD3, CD28, and CD137 antibodies and intended for ex vivo expansion/proliferation of human antigen (Ag)-specific T cells. CD3 and CD28 provide primary and co-stimulatory signals that are required for the activation and expansion of T cells. Anti-CD137 acts as an antagonist that promotes both survival and proliferation of T lymphocytes. This is especially important for memory T cells where they may lack CD28 expression.

Benefits of Dynabeads Human T-Activator CD3/CD28/CD137 beads include:

  • Easy—no need to prepare antigen-presenting cells or antigen
  • Efficient—improved expansion and increased survival of Ag-specific T-cells
  • Functional—expanded T cells maintain Ag-specific responses

Simple and fast procedure

Dynabeads Human T-Activator CD3/CD28/CD137 beads offer a simple method for expansion of CD3+, CD4+, or CD8+ antigen-specific T cells from T cell clones, cell lines, or freshly isolated Ag-specific T cells from PBMC or whole blood. Ag-specific T cells are activated by adding the beads and human recombinant IL-2 (rIL-2) and rIL7 (for CD8+ Ag-specific T cells only) to the cell culture. Optimal expansion is performed when appropriate T-cell medium is used, such as CTS OpTmizer T Cell Expansion SFM or Advanced RPMI 1640 Medium with added supplements according to the manual.

When experiencing reduced expansion or cell shrinking the T cells can be restimulated with the Dynabeads Human T-Activator CD3/CD28/CD137 beads. The beads can be removed with a DynaMag magnet and used in applications like cell:cell interaction studies, flow cytometry (after 3–4 days of culture to obtain bead release), proteomics, or nucleic acid-based assays. The magnetic beads in this product may not appear as a typical brown suspension and may not be easily visible. This is normal and does not affect performance. Be sure to thoroughly resuspend before use.

Dynabeads Human T-Activator CD3/CD28/CD137 applications

The use of Dynabeads Human T-Activator CD3/CD28/CD137 beads have also been used to expand chronic lymphocytic leukemia in vitro by activating T cells, which in turn promote the expansion of leukemic B cells (Baptista et al, 2021). In addition, the Dynabeads Human T-Activator CD3/CD28/CD137 beads have been used to understand T cell suppression in blood neoplasms (Deng, et al, 2018).

Pre-isolate human antigen-specific T cells

To obtain the most optimal Ag-specific expansion from a heterogenous T-cell population, the Ag-specific T cells can be sorted or isolated and released directly from human whole blood or from peripheral blood mononuclear cells (PBMCs) with the Dynabeads FlowComp Flexi Kit in combination with an agonistic anti-CD137 antibody.

Alternative activation products

  • For activation of naïve polyclonal human T cells, use Dynabeads Human T Activator CD3/CD28 (Cat. No. 11131D)
  • For activation of naïve polyclonal mouse T cells, use Dynabeads Mouse T Activator CD3/CD28 (Cat. No. 11456D)
  • For activation of human regulatory T cells (Tregs), use Dynabeads CD3/CD28 Treg Expander (Cat. No. 11129D)
  • For ex vivo activation and expansion of human T cells for cell-based therapy, use the aseptically manufactured CTS Dynabeads CD3/CD28 (Cat. No. 40203D)

Commercial supply

Our manufacturing sites are ISO 13485–certified. Bead characteristics and manufacturing conditions help ensure consistency and batch reproducibility, making them an ideal choice for commercial supply. If you are utilizing Dynabeads Human T-Activator CD3/CD28/CD137 beads in a commercial product or service or need a larger bulk volume of this product, please contact us at oemdynal@lifetech.com.

References

Deng M, et al., LILRB4 signalling in leukaemia cells mediates T cell suppression and tumour infiltration. Nature. 2018 Oct;562(7728):605-609.

Baptista MJ, Baskar S, Gaglione EM, Keyvanfar K, Ahn IE, Wiestner A, Sun C. Select Antitumor Cytotoxic CD8+ T Clonotypes Expand in Patients with Chronic Lymphocytic Leukemia Treated with Ibrutinib. Clin Cancer Res. 2021 Aug 15;27(16):4624-4633.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Human antigen-specific T-cells
Certifications/Compliance ISO9001 and ISO13485
Concentration 5.4 x 105 Beads/mL
Content And Storage DynaBeads Human T-Activator CD3/CD28/CD137 are supplied in PBS, pH 7.4, with 0.1% HSA. Store at 2°C to 8°C.

Description Monoclonal antibodies covalently bound to DynaBeads magnetic Beads
For Use With (Application) Activation and Expansion of T-cells
Material Magnetic Polystyrene
Product Type Antibody Coated Bead
For Use With (Equipment) DynaMag™ Magnet
Host Species Mouse
Isolation Technology Magnetic Beads
Product Line Dynabeads
Quantity 2 mL
Reactivity Human
Sample Type Pre-isolated human antigen-specific T-cells from cell lines or clones
Shipping Condition Ambient Temperature
Surface Functionality Epoxy
Target Species Human T-cells
Uniformity Monosized 4.5 μm (CV <5%)
Volume (Metric) 2 mL
Diameter (Metric) 4.5 μm
Regulatory Status For Research Use Only
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.