missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Dynabeads™ Human Treg Expander

Product Code. 10456993 Shop All Gibco Products
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10456993 2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10456993 Supplier Gibco™ Supplier No. 11129D
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Dynabeads Human Treg Expander is supplied as a suspension containing 2 x 107 Dynabeads/mL in phosphate buffered saline (PBS), pH 7.4, w/0.1% human serum albumin (HSA)

Dynabeads Human Treg Expander beads are 4.5-μm superparamagnetic beads covalently coupled with an optimized combination of anti-human CD3 and anti-human CD28 antibodies and intended for ex vivo activation and expansion/proliferation of human regulatory T cells (Tregs).

Dynabeads Human Treg Expander beads are 4.5-μm superparamagnetic beads covalently coupled with an optimized combination of anti-human CD3 and anti-human CD28 antibodies and intended for ex vivo activation and expansion/proliferation of human regulatory T cells (Tregs).

Benefits of Dynabeads Human Treg Expander beads include:
• Easy—activate and expand Tregs without using antigens or antigen-presenting cells
• Efficient—reproducible method and high expansion rate
• Retain Tregs—restimulation with beads after 2 weeks increases the expansion and avoids overgrowth of non-Tregs
• Functional—expanded Tregs retain their phenotype and suppressive function

Simple and fast procedure
The Dynabeads Human Treg Expander beads offer a simple method for the activation and expansion of Tregs. Pre-isolated Tregs are activated by adding the beads and human recombinant IL-2 (rIL-2) to the cell culture. Optimal expansion is performed when appropriate T-cell medium is used, such as CTS OpTmizer T Cell Expansion SFM (Cat. No. A1048501).

Activation of human Tregs
Short time activation: activation of Tregs with Dynabeads Human Treg Expander beads from a few hours to a few days. This procedure does not expand the cells, and proteomics expression/detection and nucleic acid assays can be performed. The cell to bead complexes cannot be used in flow cytometry in this phase.

Long-term activation/expansion: activation of Tregs with Dynabeads Human Treg Expander beads for three days to a few weeks to obtain expansion of Tregs. When experiencing reduced expansion or cell shrinking, the Tregs can be restimulated with the Dynabeads Human Treg Expander beads. The beads can be removed with a DynaMag magnet and used in cell:cell interaction studies, flow cytometry, proteomics, and nucleic acid-based assays.

Pre-isolate the human Tregs
Pure Tregs are important to obtain a good Treg population, and this can be achieved by cell sorting or pre-isolating the Tregs with our Dynabeads Regulatory CD4+CD25+ T Cell Kit (Cat. No. 11363D).

Alternative activation products, from mouse to clinic
• For ex vivo activation and expansion of human T cells for cell-based therapy, use the aseptically manufactured CTS Dynabeads CD3/CD28 (Cat. No. 40203D)
• For activation of human polyclonal T cells, use Dynabeads Human T-Activator CD3/CD28 (Cat. No. 11131D)
• For activation of human antigen-specific T cells, use Dynabeads Human T Activator CD3/CD28/CD137 (Cat. No. 11162D)
• For activation of mouse T cells, use Dynabeads Mouse T Activator CD3/CD28 (Cat. No. 11452D)

Commercial supply
Our manufacturing sites are ISO 13485–certified. Bead characteristics and manufacturing conditions help ensure consistency and batch reproducibility, making them an ideal choice for commercial supply. If you are in the process of customizing Dynabeads Human Treg Expander beads in a commercial product or service, or need a larger bulk volume of this product, please contact us at oemdynal@lifetech.com or read more on our Dynabeads OEM page.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Human regulatory T-cells
Certifications/Compliance ISO9001 and ISO13485
Clonality Monoclonal
Concentration 2 x 107 beads/mL
Content And Storage DynaBeads Human Treg Expander are supplied in PBS, pH 7.4, with 0.1% HSA. Store at 2°C to 8°C.

Description Monoclonal antibodies covalently bound to DynaBeads magnetic Beads
Color Brown
For Use With (Application) Activation and Expansion of T-cells
Material Magnetic Polystyrene
Product Type Treg Expander
For Use With (Equipment) DynaMag™ Magnet
Host Species Mouse
Isolation Technology Magnetic Beads
Product Line DynaBeads
Quantity 2 mL
Reactivity Human
Sample Type Pre-isolated human Treg cells from whole blood or PBMC
Shipping Condition Ambient temperature
Surface Functionality Epoxy
Target Species Human regulatory T-cells
Uniformity Monosized 4.5 μm (CV <5%)
Volume (Metric) 2 mL
Diameter (Metric) 4.5 μm
Regulatory Status For Research Use Only
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.