missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-basic (FGF-2/bFGF) (aa 10-155) Recombinant Protein
FGF-Basic (AA 10-155) is a bioactive protein intended for use in cell culture applications
128.00 € - 1820.00 €
Specifications
For Use With (Application) | Cell Culture |
---|---|
Gene ID (Entrez) | 2247 |
Molecular Weight (g/mol) | 17 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | FGF-Basic (AA 10-155) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10622074
|
Gibco™
PHG0024 |
10 μg |
128.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10222253
|
Gibco™
PHG0026 |
50 μg |
239.00 €
50µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10202733
|
Gibco™
PHG0021 |
100 μg |
922.00 €
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10612074
|
Gibco™
PHG0023 |
1 mg |
1820.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Human FGF-basic (FGF-2/bFGF) (aa 10-155) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
Fibroblast growth factor-basic is known as FGF-basic, bFGF, FGF 2, FGF-β, and heparin binding growth factor. FGF-basic is involved in a number of biological processes, including embryonic development and differentiation, neuron differentiation, survival, regeneration, and the proliferation of cells of mesodermal origin and many cells of neuroectodermal, ectodermal, and endodermal origin.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: proteins bioassayed with Gibco media
- High-purity, recombinant, bioactive protein: to help ensure growth factors from Life Technologies are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High biological activity: in-house activity testing of recombinant proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of BALB/3T3 cells
Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
17 kDa | |
FGF-Basic (AA 10-155) | |
Human | |
<0.1 ng/μg | |
Growth Factors & Receptors | |
Lyophilized | |
Neurobiology, Oncology, Stem Cell Research, Microbiology, Differentiation, Angiogenesis | |
95% |
2247 | |
Sequential Chromatography | |
aa 10-155 | |
Store in refrigerator (2–8°C). | |
E. coli | |
Recombinant, Ligand, Truncated | |
FGFs (Fibroblast Growth Factors) | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.