missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HEPES (Ultra Pure)

Product Code. 10115074 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10115074 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10115074 Supplier Gibco™ Supplier No. 11344041
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A zwitterionic organic chemical buffering agent commonly used in cell culture media

  • Formulated using 11g per liter of water
  • Addition of 10 to 25mM HEPES provides extra buffering capacity when cell culture requires extended periods of manipulation outside of a CO2 incubator
  • Buffering choice for many cell culture systems because it is membrane impermeable
  • Has limited effect on biochemical reactions
  • Chemically and enzymatically stable
  • Low visible and UV light absorbance

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 15 – 30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Cell Type Mammalian Cells
Form Powder
Product Type HEPES Buffer
pH 6.8 to 8.2
Quantity 1 kg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.