missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Hybridoma-SFM

Product Code. 11570416 Shop All Gibco Products
Change view
Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11570416 500 mL 500mL
10749233 1000 mL 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11570416 Supplier Gibco™ Supplier No. 12045084
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifically developed to support the growth of hybridomas for antibody production

  • Hybridoma-SFM is a serum-free, very low-protein (<20 μg/mL) medium specifically developed to support the growth of hybridomas for antibody production
  • Suitable for the culture of recombinant myeloma lines as well as traditional hybridomas, Hybridoma-SFM provides high cell densities and IgG production
  • Cholesterol-dependent cultures such as NS0 require the addition of 250X Cholesterol Lipid Concentrate
  • Hybridoma-SFM is a complete medium that is ready to use, no supplementation required
  • Store at 2°C to 8°C, protected from light

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Hybridomas
Classification Low Protein, Serum-free
Form Liquid
Product Type Serum Free Medium (SFM)
Serum Level Serum-free
With Additives Glutamine, Phenol Red
Product Line Gibco
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.