missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ LV-MAX™ Transfection Kit

Product Code. 15819081 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15819081 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15819081 Supplier Gibco™ Supplier No. A35348
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TheLV- MAX Transfection Kit is an essential component of the LV-MAX Lentiviral Production System.

The Gibco LV-MAX Transfection Kit is a vital component of the LV-MAX Lentiviral Production System, specifically designed for the transient transfection of high-density suspension-adapted Viral Production Cells (human embryonic kidney (HEK) 293-derived cells). This kit is designed to maximize lentiviral particle production, ensuring high yields and reproducibility.

Key features of the LV-MAX Transfection Kit

  • High LV yields—achieves functional titers up to 1 x 108 TU/mL (unconcentrated)
  • High-density growth—supports growth of Viral Production Cells at densities greater than 1 x 107 cells/mL for maximum vector output
  • AOF formulation—chemically defined, serum-free, and animal-origin-free medium ensures consistent performance by eliminating variability from undefined components
  • Ready-to-use—no media supplementation required; simplifies workflows and provides reliable, lot-to-lot reproducibility in lentivirus production
  • Expression system compatible—fully compatible with the LV-MAX Lentiviral Production System for seamless integration into end-to-end lentivirus manufacturing processes
  • Scalable and reproducible transfection and production—validated from 96–well plates to multi‐liter bioreactors, enabling easy scale-up without sacrificing cell yields.
  • Easy and robust culture and transfection protocols, seamlessly integrating into any transient production workflow

Kit components

  • LV-MAX Transfection Reagent—a high-efficiency, cationic, lipid-based reagent uniquely formulated for co-transfecting multiple plasmids into high-density Viral Production Cell cultures. This reagent ensures high transfection efficiency with minimal toxicity.
  • LV-MAX Supplement—an optimized, chemically defined, serum-free, protein-free, animal origin-free formulation designed to suppress cell growth during transfection. It enhances lentiviral vector production without compromising cell viability or interfering with the production process.
  • LV-MAX Enhancer—a proprietary, animal origin-free formulation that significantly boosts lentiviral vector production

The LV-MAX Transfection Kit is designed to deliver superior performance in lentiviral production workflows, providing researchers with reliable and scalable transfection solutions.

Applications

  • Gene therapy—ideal for the production of lentiviral vectors for gene therapy research and development
  • Cell therapy—supports the generation of lentiviral vectors for cell therapy applications, facilitating seamless transition from bench to clinic
  • Research and development—provides a reliable and scalable solution for viral vector production in various research settings

Available in RUO or CTS

Offered as both a Research Use Only version and a GMP-compliant Cell Therapy Systems (CTS) version (Cat. No. A4132602) to support translational and clinical-scale applications.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line Established Cell Lines
Classification Animal Origin-Free, Chemically Defined, Protein-Free, Serum-Free
Content And Storage • 50 mL LV-MAX Supplement
• 6 mL LV-MAX Transfection Reagent
• 40 mL LV-MAX Enhancer

Sufficient for 1 L LV production 

Store at 2–8°C, protect from light.
Cell Type 293-derived Cells
Sample Type Plasmid DNA
Transfection Technique Lipid-based Transfection
For Use With (Application) Transfection
Product Line LV-MAX
Product Type Transfection Kit
Quantity 1 L
Serum Compatible Yes
Shipping Condition Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.