missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MEM Vitamin Solution (100X)

Product Code. 15430554 Shop All Gibco Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15430554 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15430554 Supplier Gibco™ Supplier No. 11120052
Cell Culture Essentials
Use promo code 27976
View all promotions
 Restricted Item
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used as a growth supplement for cell culture medium, to increase cell growth and viability

  • Contains 100x the vitamins found in the standard MEM
  • For in vitro diagnostic use
  • Manufactured at a cGMP compliant facility, located in Grand Island, New York, registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards

Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Sf9 and Sf21 Cell Culture

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: -5 to -20°C
Shipping conditions: Frozen
Shelf life: 12 months from date of manufacture
Cell Type Mammalian Cells
Concentration 100 X
Form Liquid
Product Type Vitamin Supplement
Sterility Sterile-filtered
pH 5.4 to 7.7
Quantity 100 mL
Shipping Condition Dry Ice
What is the best way to handle the MEM Vitamin Solution (100X)? It comes frozen, so should I thaw it, then aliquot it and freeze the aliquots for later use?

If you're going to be using small batches of the vitamin solution at a time, it would be best to aliquot it out and store at -20 degrees C. The stability of the MEM Vitamin Solution is one year at -20 degrees C, while it is only 2 weeks at 4 degrees C.

What is the solvent used for MEM Vitamin Solution (100X)?

The solvent is distilled water.

What is Gibco MEM Vitamin Solution?

The MEM Vitamin Solution (Cat. No. 11120-052 or 11120-037 depending on your country) is a growth supplement for cell culture medium. This solution is formulated to contain 100X the vitamins found in standard MEM. Find the complete formulation here (http://www.thermofisher.com/us/en/home/technical-resources/media-formulation.163.html).

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.