missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Newborn Calf Serum, heat inactivated, New Zealand origin
GREENER_CHOICE

Product Code. 11530636
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11530636 500 mL 500mL
11520636 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11530636 Supplier Gibco™ Supplier No. 26010074

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Secure sera for cell culture with least viral risk sera from BSE-free regions.

Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
  • Sera Category: Secure
  • Origin: New Zealand
  • Collected from calves 14 days old or less

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Species Cattle/Bovine
Age Newborn (14 days or less)
Country of Origin New Zealand
Form Liquid
Format Bottle
Product Type Newborn Calf Bovine Serum
Quantity 500 mL
Serum Treatment Heat-inactivated
Product Line Gibco
Shipping Condition Frozen
Show More Show Less

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.