Learn More
Gibco™ Newborn Calf Serum, New Zealand origin

Description
Gibco sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
- Sera Category: Secure
- Origin: New Zealand
- Collected from calves 20 days old or less.
Order Info
Shipping Condition: Dry Ice
Specifications
Specifications
| Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
| Species | Cattle/Bovine |
| Age | Newborn (20 days or less) |
| Country of Origin | New Zealand |
| Form | Liquid |
| Format | Bottle |
| Product Type | Newborn Calf Bovine Serum |
| Quantity | 500 mL |
| Serum Treatment | Standard (Sterile-filtered) |
| Sterility | Sterile |
| Show More |
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.