Learn More
Beskrivelse
Sf-900 III SFM is a low-hydrolysate, serum-free, protein-free, animal origin–free insect cell culture medium optimized for the growth and maintenance of Spodoptera frugiperda (Sf9 and Sf21) cells.
- Optimized for recombinant gene expression using the baculovirus and stable insect expression systems
- Suitable for suspension and monolayer culture methods and supports growth of other lepidopteran cell lines
- Spodoptera frugiperda (Sf9) cells grown in Sf-900 III SFM achieve maximum cell densities of 10–14 x 106 cells/mL
- An improved serum-free, protein-free, animal origin–free medium designed for growth of Sf9 and other lepidopteran cell lines and production of insect viruses and recombinant proteins
- Ready to use format does not require addition of serum, glutamine, or surfactants
- Cells adapted to other commercially available serum-free media should be sequentially adapted into Sf-900 III SFM
- Reduced hydrolysate concentrations improve lot-to-lot consistency
Ordreinfo
Shipping Condition: Room temperature
Overholdelse
Gibco Sf-900 II SFM is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Tekniske data
Tekniske data
| Cell Line | Sf21, Sf9 |
| Cell Type | Insect Cell |
| Classification | Animal Origin-free, Protein-free, Serum-free |
| Form | Liquid |
| Product Type | Insect Cell Serum Free Medium (SFM) |
| Serum Level | Serum-free |
| With Additives | Glutamine |
| Product Line | Gibco, Sf-900 |
| Quantity | 1000 mL |
| Shipping Condition | Room Temperature |
| Vis mere |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.