missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Sf-900™ III SFM

Product Code. 11590446 Shop All Gibco Products
Change view
Click to view available options
Quantity:
500 mL
1000 mL
10 L
20 L
Unit Size:
10 L
1000mL
20 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11590446 10 L 10 L
11570446 500 mL 500mL
11580446 1000 mL 1000mL
15236025 20 L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11590446 Supplier Gibco™ Supplier No. 12658035
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A low-hydrolysate, serum-free, protein-free, animal origin–free insect cell culture medium

Sf-900 III SFM is a low-hydrolysate, serum-free, protein-free, animal origin–free insect cell culture medium optimized for the growth and maintenance of Spodoptera frugiperda (Sf9 and Sf21) cells.

  • Optimized for recombinant gene expression using the baculovirus and stable insect expression systems
  • Suitable for suspension and monolayer culture methods and supports growth of other lepidopteran cell lines
  • Spodoptera frugiperda (Sf9) cells grown in Sf-900 III SFM achieve maximum cell densities of 10–14 x 106 cells/mL
  • An improved serum-free, protein-free, animal origin–free medium designed for growth of Sf9 and other lepidopteran cell lines and production of insect viruses and recombinant proteins
  • Ready to use format does not require addition of serum, glutamine, or surfactants
  • Cells adapted to other commercially available serum-free media should be sequentially adapted into Sf-900 III SFM
  • Reduced hydrolysate concentrations improve lot-to-lot consistency

Order Info

Shipping Condition: Room temperature

Compliance

Gibco Sf-900 II SFM is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line Sf21, Sf9
Cell Type Insect Cell
Classification Animal Origin-free, Protein-free, Serum-free
Form Liquid
Product Type Insect Cell Serum Free Medium (SFM)
Serum Level Serum-free
With Additives Glutamine
Product Line Gibco, Sf-900
Quantity 10 L
Shipping Condition Room Temperature
Species S. frugiperda, Spodoptera frugiperda
Content And Storage Storage conditions: 2–8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 18 months from date of manufacture
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.