missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Viral Production Cells

Product Code. 17193119 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 mL
6 x 1 mL
Unit Size:
1mL
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17193119 6 x 1 mL Pack of 6
15967871 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17193119 Supplier Gibco™ Supplier No. A35827
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Viral Production Cells are a derivative of the HEK 293F cell line and are a core component of the LV-MAX Lentiviral Production System.

Gibco Viral Production Cells are a derivative of the HEK 293F cell line and are a core component of the LV-MAX Lentiviral Production System (Cat. No. A35684). Viral Production Cells have been adapted to a special chemically-defined, serum-free and protein-free LV-MAX Production Medium (A3583401). They are maintained in suspension culture and will grow to high density in LV-MAX Production Medium. Viral Production Cells are highly transfectable by LV-MAX Transfection reagent (A35348) and generate superior lentiviral production compared to polyethyleneimine (PEI)-based lentiviral production methods.

  • Suspension culture cells facilitate high-throughput screening or scale-up
  • Higher viral production levels than other suspension HEK 293 cells
  • Rapid cell growth and high culture viabilities

Frozen cells are supplied in a vial containing 1 mL of cells at 1 x 107 viable cells/mL in LV-MAX Production Medium and 10% DMSO. The cells should be thawed directly into LV-MAX Production Medium.

The cells are typically grown in flasks on a shaker platform in a humidified, CO2 cell culture incubator. However, cultures are scalable in volume from 96-deep well blocks to 2-L flasks and bioreactors. Lentiviral vector harvest is typically performed at two days post-transfection.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in liquid nitrogen until ready to use. Do not store the cells at -80°C.
Cell Line Viral Production Cell-Line
Culture Type Suspension Cell Culture
Product Type Viral Production Cells
Quantity 6 x 1 mL
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.