missing translation for 'onlineSavingsMsg'
Learn More

Giemsa Stain (Crystalline), Fisher BioReagents

Product Code. 10002764
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass Bottle
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10002764 5 g Glass Bottle 5g
10276014 10 g Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10002764 Supplier Fisher BioReagents Supplier No. BP24225
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Giemsa is a biological stain used for thin blood films to differentiate leukocytes, for staining malarial parasites in thin and thick blood films, and for staining bone marrow to show cell morphology.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51811-82-6
Molecular Formula C14H14ClN3S
Molecular Weight (g/mol) 291.80
MDL Number MFCD00012112,MFCD00081642
InChI Key NALREUIWICQLPS-UHFFFAOYSA-N
PubChem CID 13735
IUPAC Name (7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride
SMILES [Cl-].C[N+](C)=C1C=CC2=NC3=CC=C(N)C=C3SC2=C1

Specifications

Melting Point 300°C
Bio. Stain Comm. Certification Pass Test
Identification Pass Test
Formula Weight 291.8
Packaging Glass Bottle
Color Green
Quantity 5 g
Physical Form Powder
Chemical Name or Material Giemsa Stain
compliance-icons
Product Identifier
  • Giemsa Stain
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.