missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
221.00 € - 305.00 €
Specifications
| Antigen | GIF |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:500-1:2000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
GIF Polyclonal antibody specifically detects GIF in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| GIF | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| gastric intrinsic factor (vitamin B synthesis), IFIntrinsic factor, IFMHgastric intrinsic factor, INF, TCN3 | |
| Produced in rabbits immunized with purified, recombinant Human GIF (Accession#: P27352-1; Met1-Tyr417) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 2694 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title