Learn More
Cole-Parmer™ Needle Valve Adapter NPT
Product Code. 15122864 Shop All Cole-Parmer Products
Change view
Click to view available options
Type:
Needle Valve Adapter
Unit Size:
Each
Product Code. Type Unit Size
15122864 Needle Valve Adapter Each
1 options
Product Code. 15122864 Supplier Cole-Parmer™ Supplier No. 6340080
  • ONLINE EXCLUSIVE PRICE
    282.00 €
    165.18 € / Pack of 1
    Save 116.82 €
    41% Off
Estimated Shipment: 15-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Required for use with needle valve, mates with complete coupling.
Compatoble with Lab-Crest Pressure Reaction Vessels
Specifications
Type Needle Valve Adapter
For Use With (Application) Lab-Crest Pressure Reaction Vessels
Includes Rubber washer and neoprene o-ring, 1/8 in. NPT