missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ MICROMAN™ Positive-Displacement Pipets

Product Code. p-4330394 Shop All Gilson Products
Change view
Click to view available options
Volume (Metric):
1 to 10 μL
10 to 100 μL
100 to 1000 μL
20 to 50 μL
3 to 25 μL
50 to 250 μL
Model:
M10
M25
M50
M100
M250
M1000
Compatible Tips:
C10
C100
C1000
C25
C250
C50
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Model Compatible Tips unitSize
10585254 3 to 25 μL M25 C25 Each
10002082 1 to 10 μL M10 C10 Each
10561283 10 to 100 μL M100 C100 Each
10410015 20 to 50 μL M50 C50 Each
12609581 50 to 250 μL M250 C250 Each
10541113 100 to 1000 μL M1000 C1000 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10585254 Supplier Gilson™ Supplier No. F148502

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ensures accuracy and precision when pipetting problem liquids, and offers full protection against cross contamination. Gilson™ MICROMAN™ Positive-Displacement Pipets are 3 to 4 times better than an air displacement pipet when pipetting viscous or volatile liquids.

  • Piston is located inside the tip or capillary preventing pipet contamination
  • Color-coded push-button allows for easy identification of different models
  • Dual function push-button that also ejects capillary pistons
  • Comfortable PVDF handle resistant to aggressive chemicals
  • Accurately measure high vapor pressure liquids without leakage or degradation of the tip
  • M10 and M100 models are fully autoclavable

TRUSTED_SUSTAINABILITY

Specifications

Compatible Tips C25
Autoclavable Autoclavable
Model M25
Number of Channels 1
Type Positive Displacement Pipette
Volume (Metric) 3 to 25 μL
For Use With (Application) Pipetting of viscous and dense liquids
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.