missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PIPETMANTM EcoPackTM Tips

Product Code. 10570923 Shop All Gilson Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.1 to 20 μL
100 to 1000 μL
100 to 1200 μL
2 to 200 μL
20 to 300 μL
Unit Size:
Pack of 10000
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10570923 100 to 1000 μL Pack of 10000
12571087 100 to 1200 μL Pack of 10000
10071942 0.1 to 20 μL Pack of 10000
10693963 0.1 to 10 μL Pack of 10000
10614163 20 to 300 μL Pack of 10000
12507536 2 to 200 μL Pack of 10000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10570923 Supplier Gilson™ Supplier No. F161670
  • ONLINE EXCLUSIVE PRICE
    279.00 €
    214.20 € / Pack of 10000
    Save 64.80 €
    23% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ensures optimal performance of PIPETMAN pipettes across the full volume range. GilsonTM PIPETMANTM EcoPackTM Tips are fully autoclavable.

  • Optimized geometry and pure polypropylene material ensure low retention and naturally hydrophobic surface.
  • Manufactured in a cleanroom and hence no risk of contamination.
  • Full traceability from the mold to the bench guaranteed.
  • Volume graduations make it easy to visually check the volume aspirated.
  • Requires low tip-mounting force.
  • Easy identification with color-coded packaging.
  • Bulk Packs are sealed with a tamper-proof strip which can easily be opened for quick access.
  • Bulk Packs are closed using the rapid-sealing strip to prevent contamination and loss of tips.
  • Each EcoPack box contains 10,000 tips (10 bags of 1000 tips).
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 100 to 1000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Length (English) 15.59 in.
Length (Metric) 39.6 cm
Diameter (Metric) Outer .85 mm
Format Bagged
Tip Style Universal
Quantity 10000
Product Type Pipet Tips
Color Code Blue
Disposable Single-use
For Use With (Equipment) Gilson Pipetman
Graduated Yes
Product Line Pipetman EcoPack
Material Polypropylene
Graduations 300 μL; 1000 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.