missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ Safe Aspiration Station

Product Code. 11307830 Shop All Gilson Products
Change view
Click to view available options
Product Type:
Tubing Connector
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11307830 Tubing Connector Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11307830 Supplier Gilson™ Supplier No. F1077418
Estimated Shipment: 03-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Vacuum-Controlled Safe Aspiration Station with small footprint.

  • Closed, stand-alone unit designed to prevent contamination hazards
  • Compact design with adjustable speed allowing quick aspiration
  • Always ready to use by simply pressing the handle button
  • Handle accommodates different types of adaptors to aspirate from a variety of vessels
  • Convenient coupling for quick bottle emptying
  • In-line hydrophobic filter traps aerosols
  • Connect two handles to the station with the Y-shape connector
  • Motor starts only when the vacuum pressure drops
  • Level detection prevents overflow
TRUSTED_SUSTAINABILITY

Specifications

Product Type Tubing Connector
Color Clear
For Use With (Equipment) Gilson safe aspiration station
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.