missing translation for 'onlineSavingsMsg'
Learn More

Ginkgolide B, MedChemExpress

Product Code. 16484887
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484887 50 mg 50mg
16469098 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16484887 Supplier Medchem Express Supplier No. HYN0784
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ginkgolide B (BN-52021), an important active terpenoid from Ginkgo biloba leaves, is reported to increase cell viability and decrease cell apoptosis.

Ginkgolide B (BN-52021), an important active terpenoid from Ginkgo biloba leaves, is reported to increase cell viability and decrease cell apoptosis.

Specifications

Chemical Name or Material Ginkgolide B
CAS 15291-77-7
Color White
Purity Grade Notes Research
Molecular Formula C20H24O10
Quantity 50 mg
Synonym BN-52021
Solubility Information DMSO : ≥ 100 mg/mL (235.63 mM)
SMILES O[C@@H]1C(C2([C@H]3O)[C@](OC3=O)([H])O4)([C@@](O5)([H])C[C@H]2C(C)(C)C)C4(C5=O)[C@@](O)([C@@H]6C)[C@@]1([H])OC6=O
Molecular Weight (g/mol) 424.4
Formula Weight 424.4
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.