missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ginsenoside Re, MedChemExpress
Description
Ginsenoside Re (Ginsenoside B2) is an extract from Panax notoginseng. Ginsenoside Re decreases the β-amyloid protein (Aβ). Ginsenoside Re plays a role in antiinflammation through inhibition of JNK and NF-κB.
Specifications
Specifications
| Chemical Name or Material | Ginsenoside Re |
| CAS | 52286-59-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C48H82O18 |
| Quantity | 5 mg |
| Synonym | Ginsenoside B2 Panaxoside Re Sanchinoside Re |
| Solubility Information | DMSO : ≥ 50 mg/mL (52.79 mM) |
| SMILES | O[C@H]1[C@H](O)[C@@H](CO)O[C@@H](O[C@H]2C[C@]([C@@](CC[C@]3([H])[C@@](C)(O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)CC/C=C(C)/C)(C)[C@]3([H])[C@H](O)C5)(C)[C@@]5([H])[C@]6(C)[C@]2([H])C(C)(C)[C@@H](O)CC6)[C@@H]1O[C@@H]7O[C@@H](C)[C@H](O)[C@@H](O)[C@H]7O |
| Molecular Weight (g/mol) | 947.15 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?