missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ginsenoside Rh2, MedChemExpress
Description
Ginsenoside Rh2 induces the activation of caspase-8 and caspase-9. Ginsenoside Rh2 induces cancer cell apoptosis in a multi-path manner.
Specifications
Specifications
| Chemical Name or Material | Ginsenoside Rh2 |
| CAS | 78214-33-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C36H62O8 |
| Quantity | 10 mg |
| Synonym | 20(S)-Ginsenoside Rh2 20(S)-Rh2 Ginsenoside-Rh2 |
| Solubility Information | DMSO : 50 mg/mL (80.27 mM; Need ultrasonic) |
| SMILES | C[C@]([C@@](C[C@H]1O)([H])[C@]2(CC[C@@H]3O[C@]([C@@H]([C@@H](O)[C@@H]4O)O)([H])O[C@@H]4CO)C)(CC[C@@]2([H])C3(C)C)[C@]5([C@@]1([H])[C@]([C@@](C)(O)CC/C=C(C)/C)([H])CC5)C |
| Molecular Weight (g/mol) | 622.87 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?