missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Gladiator™ 16-500 Series Heavy Weight Gloves

Product Code. p-8005272
Click to view available options
Size:
7
8
9
10
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 12630772

Brand: Ansell™ 16500080

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed for light-to-medium stress glass handling. Crinkle finish provides excellent wet and dry grip on slippery surfaces but also protects the hand from sharp edges.

  • Ergonomically designed to follow the natural shape of the hand. A more natural glove shape delivers a better fit and makes for safer handling.
  • Good cushioning, good cut resistance, good comfort.
  • Crinkle finish for grip on slipper surfaces and protection from sharp edges
  • Natural rubber coating
  • Cut-and-sewn construction
  • Interlock cotton liner
  • 3/4 Dipped finish
  • Knitwrist cuff
TRUSTED_SUSTAINABILITY

Specifications

Coating Material Natural Rubber Latex
Color Green
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free No
Material Rubber Latex
Product Type Heavyweight Gloves
Size 8
Finger Style Crinkled finish
For Use With (Application) Maintenance Handling dry, wet or oily large objects Trash collecting
Glove Cuff Color Green
Glove Cuff Style Knit
Liner Material Interlock Cotton
Lining Lined
Quantity 12 Pairs
Resistant To Good cushioning, good cut resistance
Series 16-500
Length (English) 9.1
Length (Metric) 230
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.