missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- A drip free pour ring and autoclavable polypropylene (PP) screw cap enable sterilization of media and reagents.
- Globe Glass™ amber media bottles are a great choice for mixing, storing, and transport of light sensitive solutions and media.
- Available in sizes 50 mL to 2000 mL.
- ASTM E438, Type I, Class A, Borosilicate 3.3 Glass
- Amber color reduces light transmission
- GlassGuard™ packaging system protects against transit damage
- GL32 polypropylene (PP) screw cap on size 50 mL
- GL45 polypropylene (PP) screw caps on sizes 100 mL to 2 L
- Pouring ring for drip-free liquid transfer
- High resistance to chemicals
- Dual scale metric graduations
- Durable white enamel printing
- Large white marking area
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Color | Amber |
| Material | Borosilicate Glass |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | Yes |
| Mouth | Threaded |
| Closure Size | GL45 |
| Autoclavable | Autoclavable |
| Capacity (English) | 16.91 oz. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?