missing translation for 'onlineSavingsMsg'
Learn More

Hecht Karl™ Glass Beads

Product Code. 10449411
Change view
Click to view available options
Diameter (Metric):
3 mm
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
10449411 3 mm 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10449411 Supplier Hecht Karl™ Supplier No. 41401003
  • ONLINE EXCLUSIVE PRICE
    44.21 €
    30.46 € / 1kg
    Save 13.75 €
    31% Off
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.